Fmoc-N-Me-Nle-OH 96%

Supplier: Thermo Scientific Chemicals

Synonyms: N-Fmoc-N-methyl-L-norleucine

H63992-03 H63992-06
AAH63992-03EA 46.59 USD
AAH63992-03 AAH63992-06
Fmoc-N-Me-Nle-OH 96%
Fmoc-N-Me-Nle-OH
Formula: C₂₂H₂₅NO₄
MW: 367.445 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00235878
CAS Number: 112883-42-8

Order Now

Specification Test Results

Assay 96%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR