2,4,6,8-Tetrachloroquinazoline 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H63968-MD
H63968-03
AAH63968-03EA
751.47
USD
AAH63968-03
AAH63968-MD
2,4,6,8-Tetrachloroquinazoline 97%
2,4,6,8-Tetrachloroquinazoline
Formula:
C₈H₂Cl₄N₂ Storage Temperature: Ambient |
MDL Number:
MFCD03844695 CAS Number: 54185-42-1 |
Specification Test Results
Assay | 97% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...