Fmoc-L-Arg(Mtr)-OH 95%

Warning

Synonyms: Nα-Fmoc-Nomega-(4-methoxy-2,3,6-trimethylphenylsulphonyl)-L-arginineFmoc-Arg(Mtr)-OH

H63907-14 H63907-22 H63907-06
AAH63907-14EA 156.51 USD
AAH63907-14 AAH63907-22 AAH63907-06
Fmoc-L-Arg(Mtr)-OH 95%
Fmoc-L-Arg(Mtr)-OH
Formula: C₃₁H₃₆N₄O₇S
MW: 608.71 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00065616
CAS Number: 98930-01-9

Order Now

Specification Test Results

Assay 95%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR