H-Cys(pMeOBzl)-OH tech. 80%, Technical Grade
Synonyms:
H-Cys(Mob)-OH
H63894-06
H63894-14
AAH63894-06EA
54.46
USD
AAH63894-06
AAH63894-14
H-Cys(pMeOBzl)-OH tech. 80%, Technical Grade
H-Cys(pMeOBzl)-OH
Formula:
C₁₁H₁₅NO₃S MW: 241.305 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00037207 CAS Number: 2544-31-2 |
Specification Test Results
Assay | 80% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...