Fmoc-D-Phg-OH 98%

Supplier: Thermo Scientific Chemicals

Synonyms: N-Fmoc-D-2-phenylglycineFmoc-D-Phg-OHN-Fmoc-D-α-phenylglycineN-[(9H-Fluoren-9-ylmethoxy)carbonyl]-D-2-phenylglycine

H63823-14 H63823-03 H63823-06
AAH63823-06EA 84.34 USD
AAH63823-06 AAH63823-14 AAH63823-03
Fmoc-D-Phg-OH 98%
Fmoc-D-Phg-OH
Formula: C₂₃H₁₉NO₄
MW: 373.408 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00190891
CAS Number: 111524-95-9

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR