Fmoc-Leu-OPfp 98%

Synonyms: N-Fmoc-L-leucine pentafluorophenyl ester

H63820-06 H63820-03
AAH63820-03EA 28.34 USD
AAH63820-03 AAH63820-06
Fmoc-Leu-OPfp 98%
Fmoc-Leu-OPfp
Formula: C₂₇H₂₂F₅NO₄
Storage Temperature: Ambient
MDL Number: MFCD00065658
CAS Number: 86060-88-0

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR