3-Bromo-5-(trifluoromethyl)benzoic acid 97%

Warning

H63776-06 H63776-22 H63776-14
AAH63776-14EA 145.41 USD
AAH63776-14 AAH63776-22 AAH63776-06
3-Bromo-5-(trifluoromethyl)benzoic acid 97%
3-Bromo-5-(trifluoromethyl)benzoic acid
Formula: C₈H₄BrF₃O₂
MW: 269.02 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03412186
CAS Number: 328-67-6

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR