4-Chloro-2-fluoro-5-nitrobenzoic acid 98%
|
Warning
|
H63760-03
H63760-06
H63760-MD
AAH63760-MDEA
16.07
USD
AAH63760-MD
AAH63760-03
AAH63760-06
4-Chloro-2-fluoro-5-nitrobenzoic acid 98%
4-Chloro-2-fluoro-5-nitrobenzoic acid
Formula:
C₇H₃ClFNO MW: 219.55 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD04115725 CAS Number: 35112-05-1 |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...