(S)-2-(((Benzyloxy)carbonyl)amino)-2-cyclohexylacetic acid 95%

Synonyms: Z-cyclohexyl-Gly-OH

H63723-06 H63723-03
AAH63723-06EA 165.39 USD
AAH63723-06 AAH63723-03
(S)-2-(((Benzyloxy)carbonyl)amino)-2-cyclohexylacetic acid 95%
(S)-2-(((Benzyloxy)carbonyl)amino)-2-cyclohexylacetic acid
Formula: C₁₆H₂₁NO₄
MW: 291.347 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00191083
CAS Number: 69901-75-3

Order Now

Specification Test Results

Assay 95%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR