L-Aspartic acid 4-cyclohexyl ester 95%

Synonyms: H-Asp(OcHex)-OH

H63707-22 H63707-06 H63707-14
AAH63707-06EA 34.13 USD
AAH63707-06 AAH63707-22 AAH63707-14
L-Aspartic acid 4-cyclohexyl ester 95%
L-Aspartic acid 4-cyclohexyl ester
Formula: C₁₀H₁₇NO₄
MW: 215.249 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00190762
CAS Number: 112259-66-2

Order Now

Specification Test Results

Assay 95%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR