2-Amino-4-[2-(boc-amino)ethyl]thiazole 97%

Supplier: Thermo Scientific Chemicals
Warning

H63689-MD H63689-03
AAH63689-03EA 785.99 USD
AAH63689-03 AAH63689-MD
2-Amino-4-[2-(boc-amino)ethyl]thiazole 97%
2-Amino-4-[2-(boc-amino)ethyl]thiazole
Formula: C₁₀H₁₇N₃O₂S
Storage Temperature: Ambient
MDL Number: MFCD09998237
CAS Number: 956018-34-1

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR