3-(2-Thienyl)-D-alanine 95%
|
Warning
|
Synonyms:
H-β-(2-Thienyl)-D-Ala-OH
H63681-03
H63681-MD
AAH63681-03EA
318.57
USD
AAH63681-03
AAH63681-MD
3-(2-Thienyl)-D-alanine 95%
3-(2-Thienyl)-D-alanine
Formula:
C₇H₉NO₂S MW: 171.214 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00077166 CAS Number: 62561-76-6 |
Specification Test Results
Assay | 95% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...