Boc-D-Chg-OH 95%

Synonyms: Boc-cyclohexyl-D-Gly-OH

H63673-06 H63673-03
AAH63673-03EA 132.44 USD
AAH63673-03 AAH63673-06
Boc-D-Chg-OH 95%
Boc-D-Chg-OH
Formula: C₁₃H₂₃NO₄
MW: 257.33 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00133629
CAS Number: 70491-05-3

Order Now

Specification Test Results

Assay 95%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR