Methyl-6-bromo-1-methyl-1H-indole-2-carboxylate 97%

Supplier: Thermo Scientific Chemicals

Synonyms: Methyl 6-bromo-1-methylindole-2-carboxylate

H63655-MD H63655-03 H63655-06
AAH63655-03EA 103.99 USD
AAH63655-03 AAH63655-06 AAH63655-MD
Methyl-6-bromo-1-methyl-1H-indole-2-carboxylate 97%
Methyl-6-bromo-1-methyl-1H-indole-2-carboxylate
Formula: C₁₁H₁₀BrNO₂
MW: 268.11 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07364734
CAS Number: 680569-18-0

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR