3-Bromo-4-fluorophenylacetic acid 96%

Warning

H63622-14 H63622-06 H63622-03
AAH63622-06EA 50.82 USD
AAH63622-06 AAH63622-14 AAH63622-03
3-Bromo-4-fluorophenylacetic acid 96%
3-Bromo-4-fluorophenylacetic acid
Formula: C₈H₆BrFO₂
MW: 233.04 g/mol
Melting Pt: 65…69 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD07368122
CAS Number: 194019-11-9

Order Now

Specification Test Results

Assay 96%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR