Fmoc-N-Me-Ser(tBu)-OH 97%
Supplier: Thermo Scientific Chemicals
Synonyms:
N-Fmoc-O-tert-butyl-N-methyl-L-serine
H63614-MD
H63614-03
H63614-06
AAH63614-MDEA
41.84
USD
AAH63614-MD
AAH63614-03
AAH63614-06
Fmoc-N-Me-Ser(tBu)-OH 97%
Fmoc-N-Me-Ser(tBu)-OH
Formula:
C₂₃H₂₇NO₅ Melting Pt: 213…218 °C Storage Temperature: Ambient |
MDL Number:
MFCD02094430 CAS Number: 197632-77-2 |
Specification Test Results
Assay | 97% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...