2,3,5-Trichloroisonicotinic acid 95%

Warning

Synonyms: 2,3,5-Trichloropyridine-4-carboxylic acid

H63588-03 H63588-MD H63588-06
AAH63588-06EA 237.54 USD
AAH63588-06 AAH63588-03 AAH63588-MD
2,3,5-Trichloroisonicotinic acid 95%
2,3,5-Trichloroisonicotinic acid
Formula: C6H2Cl3NO2
MW: 226.44 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11044390
CAS Number: 406676-18-4

Order Now

Specification Test Results

Assay 95%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR