Methyl-1-methyl-5-oxopyrrolidine-3-carboxylate 97%
|
Warning
|
Synonyms:
Methyl 1-Methyl-2-oxopyrrolidine-4-carboxylate
H63577-MD
H63577-06
H63577-03
AAH63577-MDEA
56.73
USD
AAH63577-MD
AAH63577-03
AAH63577-06
Methyl-1-methyl-5-oxopyrrolidine-3-carboxylate 97%
Methyl-1-methyl-5-oxopyrrolidine-3-carboxylate
Formula:
C₇H₁₁NO₃ MW: 157.169 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00015898 CAS Number: 59857-86-2 |
Specification Test Results
Assay | 97% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...