Bis[(2R,5R)-1-(2-[(2R,5R)-2,5-dimethyl-1-phospholanyl]phenyl)-2,5-dimethylphospholane 1-oxide]copper(I) triflate 97%
|
Warning
|
H63492-ME
H63492-MC
AAH63492-MCEA
72.59
USD
AAH63492-MC
AAH63492-ME
Bis[(2R,5R)-1-(2-[(2R,5R)-2,5-dimethyl-1-phospholanyl]phenyl)-2,5-dimethylphospholane 1-oxide]copper(I) triflate 97%
Bis[(2R,5R)-1-(2-[(2R,5R)-2,5-dimethyl-1-phospholanyl]phenyl)-2,5-dimethylphospholane 1-oxide]copper(I) triflate
Formula:
C₃₇H₅₆CuF₃O₅P₄S Storage Temperature: Ambient |
MDL Number:
MFCD17018797 CAS Number: 874013-62-4 |
Specification Test Results
Assay | 97% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...