Z-Tyr(Bzl)-OH 95%

Synonyms: N-Benzyloxycarbonyl-O-benzyl-L-tyrosine

H63487-03 H63487-06
AAH63487-06EA 183.62 USD
AAH63487-06 AAH63487-03
Z-Tyr(Bzl)-OH 95%
Z-Tyr(Bzl)-OH
Formula: C₂₄H₂₃NO₅
Storage Temperature: Ambient
MDL Number: MFCD00037819
CAS Number: 16677-29-5

Order Now

Specification Test Results

Assay 95%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR