L-Tyrosine benzyl ester-p-toluenesulfonate 98%

Synonyms: H-Tyr-OBzl.Tos-OH

H63361-22 H63361-14
AAH63361-14EA 70 USD
AAH63361-14 AAH63361-22
L-Tyrosine benzyl ester-p-toluenesulfonate 98%
L-Tyrosine benzyl ester-p-toluenesulfonate
Formula: C₁₆H₁₇NO₃
MW: 443.51 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00038959
CAS Number: 53587-11-4
EINECS: 258-650-8

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR