2-Chloro-5-(trifluoromethyl)-4-iodopyridine 90%, Technical Grade

Warning

Synonyms: 2-Chloro-4-iodo-5-(trifluoromethyl)pyridine

H63345-03 H63345-MD H63345-06
AAH63345-03EA 99.04 USD
AAH63345-03 AAH63345-06 AAH63345-MD
2-Chloro-5-(trifluoromethyl)-4-iodopyridine 90%, Technical Grade
2-Chloro-5-(trifluoromethyl)-4-iodopyridine
Formula: C6H2ClF3IN
MW: 307.44 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01862099
CAS Number: 505084-55-9

Order Now

Specification Test Results

Assay 90%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR