8-Chloro[1,2,4]triazolo[4,3-a]pyrazine 97%
|
Warning
|
H63343-MD
H63343-06
H63343-03
AAH63343-MDEA
32.25
USD
AAH63343-MD
AAH63343-06
AAH63343-03
8-Chloro[1,2,4]triazolo[4,3-a]pyrazine 97%
8-Chloro[1,2,4]triazolo[4,3-a]pyrazine
Formula:
C5H3ClN4 MW: 154.56 g/mol Melting Pt: 215…220 °C Storage Temperature: Ambient |
MDL Number:
MFCD08272086 CAS Number: 68774-77-6 |
Specification Test Results
Assay | 97% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...