4,5-Dibromo-2-fluorotoluene 98%

Warning

Synonyms: 1,2-Dibromo-4-fluoro-5-methylbenzene

H63321-03 H63321-06 H63321-MD
AAH63321-03EA 59.96 USD
AAH63321-03 AAH63321-06 AAH63321-MD
4,5-Dibromo-2-fluorotoluene 98%
4,5-Dibromo-2-fluorotoluene
Formula: C₇H₅Br₂F
MW: 267.92 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00143234
CAS Number: 202982-77-2

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR