Ethyl-4-acetylbenzoate 98%

Warning

Synonyms: Ethyl 4-acetylbenzoate

H63258-03 H63258-06
AAH63258-06EA 216.97 USD
AAH63258-06 AAH63258-03
Ethyl-4-acetylbenzoate 98%
Ethyl-4-acetylbenzoate
Formula: C₁₁H₁₂O₃
MW: 192.214 g/mol
Melting Pt: 55…57 °C
Storage Temperature: Ambient
MDL Number: MFCD00013241
CAS Number: 38430-55-6

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR