N-Methyl-L-proline 98%

Synonyms: N-Me-Pro-OH

H63241-MD H63241-03
AAH63241-MDEA 141.9 USD
AAH63241-MD AAH63241-03
N-Methyl-L-proline 98%
N-Methyl-L-proline
Formula: C₆H₁₁NO₂
MW: 129.159 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00011565
CAS Number: 475-11-6

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR