N-Fmoc-L-homoarginine 96%

Synonyms: N6-(Aminoiminomethyl)-N2-Fmoc-L-lysineFmoc-HoArg-OH(S)-2-(Fmoc-amino)-6-guanidinohexanoic acidN6-(Aminoiminomethyl)-N2-[(9H-fluoren-9-ylmethoxy)carbonyl]-L-lysineFmoc-Homoarg-OhN-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-homoarginine

H63228-03 H63228-MD H63228-06
AAH63228-06EA 475.08 USD
AAH63228-06 AAH63228-MD AAH63228-03
N-Fmoc-L-homoarginine 96%
N-Fmoc-L-homoarginine
Formula: C₂₂H₂₆N₄O₄
MW: 410.474 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01861360
CAS Number: 776277-76-0

Order Now

Specification Test Results

Assay 96%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR