2-Chloro-4-[2-(4-fluorophenyl)ethyl]-6-methylpyrimidine 97%

Warning

H63212-MD H63212-03
AAH63212-MDEA 88.87 USD
AAH63212-MD AAH63212-03
2-Chloro-4-[2-(4-fluorophenyl)ethyl]-6-methylpyrimidine 97%
2-Chloro-4-[2-(4-fluorophenyl)ethyl]-6-methylpyrimidine
Formula: C₁₃H₁₂ClFN₂
Storage Temperature: Ambient
MDL Number: MFCD04974533
CAS Number: 372183-70-5

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR