N-Acetyl-DL-phenylalanine 95%

Synonyms: Ac-DL-Phe-OH

H63125-22 H63125-14
AAH63125-22EA 120.99 USD
AAH63125-22 AAH63125-14
N-Acetyl-DL-phenylalanine 95%
N-Acetyl-DL-phenylalanine
Formula: C₁₁H₁₃NO₃
MW: 207.229 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00063157
CAS Number: 2901-75-9
EINECS: 220-793-9

Order Now

Specification Test Results

Assay 95%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR