(4-nitro-1H-indol-3-yl)methanol 97%

Warning

Synonyms: 3-Hydroxymethyl-4-nitroindole

H63120-MD H63120-03
AAH63120-MDEA 122.18 USD
AAH63120-MD AAH63120-03
(4-nitro-1H-indol-3-yl)methanol 97%
(4-nitro-1H-indol-3-yl)methanol
Formula: C₉H₈N₂O₃
Storage Temperature: Ambient
MDL Number: MFCD04973980
CAS Number: 124549-51-5

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR