3-Fluoro-2-nitrobenzoic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

H63051-MD H63051-03 H63051-06
AAH63051-06EA 386.99 USD
AAH63051-06 AAH63051-MD AAH63051-03
3-Fluoro-2-nitrobenzoic acid 98%
3-Fluoro-2-nitrobenzoic acid
Formula: C₇H₄FNO₄
MW: 185.11 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07368342
CAS Number: 1000339-51-4

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR