6-Nitro-1H-indole-2-carboxylic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 6-Nitroindole-2-carboxylic acid

H63032-06 H63032-MD H63032-03
AAH63032-03EA 200.56 USD
AAH63032-03 AAH63032-MD AAH63032-06
6-Nitro-1H-indole-2-carboxylic acid 97%
6-Nitro-1H-indole-2-carboxylic acid
Formula: C₉H₆N₂O₄
Storage Temperature: Ambient
MDL Number: MFCD02664466
CAS Number: 10242-00-9

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR