Boc-Lys(boc)-ONP 98%

Synonyms: N2,N6-Di-Boc-L-Lysine 4-Nitrophenyl EsterNα,Nepsilon-Di-Boc-L-lysine 4-nitrophenyl ester

H62949-06 H62949-14
AAH62949-06EA 51.73 USD
AAH62949-06 AAH62949-14
Boc-Lys(boc)-ONP 98%
Boc-Lys(boc)-ONP
Formula: C₂₂H₃₃N₃O₈
MW: 467.519 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03940100
CAS Number: 2592-19-0
EINECS: 219-988-1

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR