Ethyl-1-(hydroxymethyl)cyclopentanecarboxylate 97%

Supplier: Thermo Scientific Chemicals

H62804-MD H62804-03
AAH62804-03EA 328.99 USD
AAH62804-03 AAH62804-MD
Ethyl-1-(hydroxymethyl)cyclopentanecarboxylate 97%
Ethyl-1-(hydroxymethyl)cyclopentanecarboxylate
Formula: C₉H₁₆O₃
Storage Temperature: Ambient
MDL Number: MFCD12022629
CAS Number: 1075-82-7

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR