L-phenylalanine benzyl ester-p-toluenesulfonate 95%

Synonyms: H-Phe-OBzl·p-tosylate

H62700-06 H62700-14
AAH62700-06EA 33.99 USD
AAH62700-06 AAH62700-14
L-phenylalanine benzyl ester-p-toluenesulfonate 95%
L-phenylalanine benzyl ester-p-toluenesulfonate
Formula: C₂₃H₂₅NO₅S
MW: 427.515 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00035105
CAS Number: 1738-78-9
EINECS: 217-096-7

Order Now

Specification Test Results

Assay 95%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR