(R)-1-tert-Butyl-2-methyl pyrrolidine-1,2-dicarboxylate 95%

Synonyms: Boc-D-Pro-OMeN-(tert-Butoxycarbonyl)-D-proline methyll esterN-Boc-D-proline methyll esterN-Boc-D-proline methyl ester

H62672-03 H62672-MD H62672-06
AAH62672-03EA 85.4 USD
AAH62672-03 AAH62672-MD AAH62672-06
(R)-1-tert-Butyl-2-methyl pyrrolidine-1,2-dicarboxylate 95%
(R)-1-tert-Butyl-2-methyl pyrrolidine-1,2-dicarboxylate
Formula: C11H19NO4
MW: 229.27 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00190828
CAS Number: 73323-65-6

Order Now

Specification Test Results

Assay 95%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR