N-Boc-1-benzyloxymethyl-L-histidine 97%

Synonyms: Boc-His(Bom)-OHN-[(1,1-Dimethylethoxy)carbonyl]-1-[(phenylmethoxy)methyl]- L-histidine

H62663-MD H62663-03 H62663-06
AAH62663-03EA 53.63 USD
AAH62663-03 AAH62663-MD AAH62663-06
N-Boc-1-benzyloxymethyl-L-histidine 97%
N-Boc-1-benzyloxymethyl-L-histidine
Formula: C₁₉H₂₅N₃O₅
MW: 375.42 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00038627
CAS Number: 83468-83-1

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR