N-Boc-3-trifluoromethyl-L-phenylalanine 98%

Synonyms: Boc-Phe(3-CF3)-OH(S)-2-((tert-Butoxycarbonyl)amino)-3-(3-(trifluoromethyl)phenyl)propanoic acid

H62637-06 H62637-03
AAH62637-03EA 295.07 USD
AAH62637-03 AAH62637-06
N-Boc-3-trifluoromethyl-L-phenylalanine 98%
N-Boc-3-trifluoromethyl-L-phenylalanine
Formula: C₁₅H₁₈F₃NO₄
MW: 333.307 g/mol
Melting Pt: 135…138 °C
Storage Temperature: Ambient
MDL Number: MFCD00672524
CAS Number: 142995-31-1

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR