1-[3,5-Bis(trifluoromethyl)phenyl]-3-(4-fluorophenyl)urea 97%

Warning

H62583-MD H62583-03 H62583-06
AAH62583-MDEA 88.17 USD
AAH62583-MD AAH62583-06 AAH62583-03
1-[3,5-Bis(trifluoromethyl)phenyl]-3-(4-fluorophenyl)urea 97%
1-[3,5-Bis(trifluoromethyl)phenyl]-3-(4-fluorophenyl)urea
Formula: C₁₅H₉F₇N₂O
Storage Temperature: Ambient
MDL Number: MFCD04069619
CAS Number: 978-65-4

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR