cis-3-(Benzyloxymethyl)cyclobutanol 97%

Supplier: Thermo Scientific Chemicals

H62579-03 H62579-MD
AAH62579-03EA 630.99 USD
AAH62579-03 AAH62579-MD
cis-3-(Benzyloxymethyl)cyclobutanol 97%
cis-3-(Benzyloxymethyl)cyclobutanol
Formula: C₁₂H₁₆O₂
MW: 192.258 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06657676
CAS Number: 172324-68-4

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR