cis-1-(Boc-amino)-4-(2-hydroxyethyl)cyclohexane 97%

Supplier: Thermo Scientific Chemicals

H62557-03 H62557-MD
AAH62557-MDEA 236.9 USD
AAH62557-MD AAH62557-03
cis-1-(Boc-amino)-4-(2-hydroxyethyl)cyclohexane 97%
cis-1-(Boc-amino)-4-(2-hydroxyethyl)cyclohexane
Formula: C₁₃H₂₅NO₃
MW: 243.347 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03844625
CAS Number: 1069120-19-9

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR