Boc-D-leucinol 98%

Supplier: Thermo Scientific Chemicals

Synonyms: N-Boc-D-leucinol

H62498-03
AAH62498-03EA 69.69 USD
AAH62498-03
Boc-D-leucinol 98%
Boc-D-leucinol
Formula: C₁₁H₂₃NO₃
MW: 217.309 g/mol
Boiling Pt: 213 °C
Density: 0.983
Flash Pt: 110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00235931
CAS Number: 106930-51-2

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR