(S)-2-((tert-Butoxycarbonyl)amino)-2-cyclohexylacetic acid 97%

Synonyms: Boc-cyclohexyl-Gly-OHBoc-Chg-OHN-Boc-L-2-cyclohexylglycine

H62466-06 H62466-03
AAH62466-06EA 239.76 USD
AAH62466-06 AAH62466-03
(S)-2-((tert-Butoxycarbonyl)amino)-2-cyclohexylacetic acid 97%
(S)-2-((tert-Butoxycarbonyl)amino)-2-cyclohexylacetic acid
Formula: C13H23NO4
MW: 257.3 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00133638
CAS Number: 109183-71-3

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR