trans-4-(Benzyloxycarbonylamino)cyclohexylamine 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Benzyl trans-4-(aminomethyl)cyclohexylcarbamate hydrochloride

H62444-06 H62444-MD H62444-03
AAH62444-MDEA 24 USD
AAH62444-MD AAH62444-06 AAH62444-03
trans-4-(Benzyloxycarbonylamino)cyclohexylamine 97%
trans-4-(Benzyloxycarbonylamino)cyclohexylamine
Formula: C₁₄H₂₀N₂O₂
MW: 248.326 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03839901
CAS Number: 149423-77-8

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR