D-Alanine benzyl ester-p-toluenesulfonate 98%

Synonyms: H-D-Ala-OBzl.Tos-OH

H62412-06 H62412-14
AAH62412-06EA 157.62 USD
AAH62412-06 AAH62412-14
D-Alanine benzyl ester-p-toluenesulfonate 98%
D-Alanine benzyl ester-p-toluenesulfonate
Formula: C₁₀H₁₃NO₂
MW: 351.42 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00039092
CAS Number: 41036-32-2
EINECS: 255-185-2

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR