trans-1-(Boc-amino)-4-(N-methoxy-N-methylcarbamoyl)cyclohexane 97%
Supplier: Thermo Scientific Chemicals
H62397-03
H62397-06
H62397-MD
AAH62397-03EA
218.99
USD
AAH62397-03
AAH62397-MD
AAH62397-06
trans-1-(Boc-amino)-4-(N-methoxy-N-methylcarbamoyl)cyclohexane 97%
trans-1-(Boc-amino)-4-(N-methoxy-N-methylcarbamoyl)cyclohexane
Formula:
C₁₄H₂₆N₂O₄ MW: 286.372 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD08436075 CAS Number: 400898-92-2 |
Specification Test Results
Assay | 97% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...