N-Boc-sarcosine methyl ester 97%

Supplier: Thermo Scientific Chemicals

Synonyms: Boc-Gly-OmeN-Boc-N-methylglycine methyl ester

H62304-06 H62304-MD H62304-03
AAH62304-MDEA 34.38 USD
AAH62304-MD AAH62304-06 AAH62304-03
N-Boc-sarcosine methyl ester 97%
N-Boc-sarcosine methyl ester
Formula: C₉H₁₇NO₄
MW: 203.24 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04973115
CAS Number: 42492-57-9

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR