N-Boc-3-(2-furyl)-L-alanine dicyclohexylammonium salt 98%

Synonyms: Dicyclohexylamine (S)-2-((tert-butoxycarbonyl)amino)-3-(furan-2-yl)propanoate

H62258-MD H62258-03 H62258-06
AAH62258-06EA 579.42 USD
AAH62258-06 AAH62258-MD AAH62258-03
N-Boc-3-(2-furyl)-L-alanine dicyclohexylammonium salt 98%
N-Boc-3-(2-furyl)-L-alanine dicyclohexylammonium salt
Formula: C₂₄H₄₀N₂O₅
MW: 436.593 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01860894
CAS Number: 331730-08-6

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR