Fmoc-Lys(Tfa)-OH 95%

Synonyms: Nα-Fmoc-Nepsilon-trifluoracetyl-L-lysine

H62243-14 H62243-03 H62243-06
AAH62243-06EA 120.59 USD
AAH62243-06 AAH62243-14 AAH62243-03
Fmoc-Lys(Tfa)-OH 95%
Fmoc-Lys(Tfa)-OH
Formula: C₂₃H₂₃F₃N₂O₅
Melting Pt: 170…174 °C
Storage Temperature: Ambient
MDL Number: MFCD00153360
CAS Number: 76265-69-5

Order Now

Specification Test Results

Assay 95%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR