trans-3-(Boc-amino)cyclohexanecarboxylic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

H62207-MD H62207-03
AAH62207-MDEA 46.77 USD
AAH62207-MD AAH62207-03
trans-3-(Boc-amino)cyclohexanecarboxylic acid 97%
trans-3-(Boc-amino)cyclohexanecarboxylic acid
Formula: C₁₂H₂₁NO₄
MW: 243.303 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03788640
CAS Number: 218772-92-0

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR